missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4orf42 Monoclonal Antibody (OTI2C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503479S
This item is not returnable.
View return policy
Description
C4orf42 Monoclonal antibody specifically detects C4orf42 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
| C4orf42 | |
| Monoclonal | |
| 4.03 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q07343 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5142 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry | |
| OTI2C6 | |
| Unconjugated | |
| PDE4B | |
| C4orf42; CTBP1 antisense RNA 2 (head to head); CTBP1-AS1; CTBP1-AS2 | |
| Full length human recombinant protein of human PDE4B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction