missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4orf42 Monoclonal Antibody (OTI4F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503514
This item is not returnable.
View return policy
Description
C4orf42 (CTBP1-DT, CTBP1 Divergent Transcript) is an RNA Gene, and is affiliated with the non-coding RNA class.Specifications
| C4orf42 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| C4orf42, CTBP1-AS1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 92070 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot | |
| OTI4F2 | |
| Unconjugated | |
| CTBP1-DT | |
| CTBP1-DT | |
| Human recombit protein fragment corresponding to amino acids 1-186 of human C4orf42 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction