missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C5AR1 Monoclonal Antibody (S5/1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516937
This item is not returnable.
View return policy
Description
Predicted to react with mustelid based on sequence homology. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD88, also known as C5aR1, is a C5a Anaphylatoxin Receptor that mediates the effects of one of the most potent chemotactic agents known. C5AR1 has been implicated in asthma, and its expression has been reported in myeloid blood cells, brain, liver, lung, spleen, heart, kidney, and intestinal tract. CD88 is the receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. Functionally, CD88 stimulates chemotaxis, granule enzyme release and superoxide anion production. CD88 is also a component of the classical pathway of the complement cascade; it may be involved with tissue injury associated with ischemia or sepsis and it may be a component of the membrane attack complex. Diseases associated with CD88 include Mannose-Binding Lectin Deficiency and Systemic Lupus Erythematosus.
Specifications
| C5AR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P21730, Q9TUE1 | |
| C5AR1 | |
| C5aR - peptide: Met1 - Asn31. | |
| 200 μg | |
| Primary | |
| Human, Rabbit, Bovine, Mink, Ferret | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin), Western Blot | |
| S5/1 | |
| Unconjugated | |
| C5AR1 | |
| anaphylatoxin C5a receptor; C5A; C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5AR; C5a-R; C5AR1; C5r1; Cd88; CD88 antigen; complement C5a receptor; complement C5a receptor 1; complement component 5 receptor 1; complement component 5, receptor 1; complement component 5a receptor 1; D7Msu1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 100352978, 101692871, 493645, 728 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction