missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C5AR1 Recombinant Mouse Monoclonal Antibody (32-G1)
GREENER_CHOICE

Mouse Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA548289

Product Code. 17951041

  • € 560.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specificity: The 32-G1 antibody recognizes C5aR which has its serine 334 phosphorylated. The mAb 32-G1 reacted specifically with a C5aR peptide that contained phosphoserine 334 and also reacted with C5aRs that were isolated from C5a-stimulated, transfected RBL-C5aR cells. It did not recognize a receptor mutant (C5aR-S--) with alanine replacement of all six C-terminal serine phosphorylation sites. Antibody 32-G1 specifically reacts with C5aR phosphoserine 334, independent of the phosphorylation state of the neighboring serine residues. Studies with Bt2cAMP-treated U937 cells show that 32-G1 specifically reacts with ligand-activated C5aR at endogenous receptor expression levels but only weakly recognizes C5aR isolated from unstimulated cells.

CD88, also known as C5aR1, is a C5a Anaphylatoxin Receptor that mediates the effects of one of the most potent chemotactic agents known. C5AR1 has been implicated in asthma, and its expression has been reported in myeloid blood cells, brain, liver, lung, spleen, heart, kidney, and intestinal tract. CD88 is the receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. Functionally, CD88 stimulates chemotaxis, granule enzyme release and superoxide anion production. CD88 is also a component of the classical pathway of the complement cascade; it may be involved with tissue injury associated with ischemia or sepsis and it may be a component of the membrane attack complex. Diseases associated with CD88 include Mannose-Binding Lectin Deficiency and Systemic Lupus Erythematosus.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C5AR1
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P21730
C5AR1
This antibody was raised by immunizing mice a synthetic C5aR C-terminal phosphopeptide that contained phosphoserine instead of serine at position 334 and also at the neighboring phosphorylation sites Ser-332 and Ser-338.
200 μg
Primary
Human
Antibody
IgG1 κ
Western Blot, Immunocytochemistry
32-G1
Unconjugated
C5AR1
anaphylatoxin C5a receptor; C5A; C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5AR; C5a-R; C5AR1; C5r1; Cd88; CD88 antigen; complement C5a receptor; complement C5a receptor 1; complement component 5 receptor 1; complement component 5, receptor 1; complement component 5a receptor 1; D7Msu1
Mouse
Protein A
RUO
728
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.