missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C5orf35 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15848333
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15848333 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15848333 Supplier Invitrogen™ Supplier No. PA569924

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 93%.

SETD9 gene ontology annotations related to this gene include regulation of signal transduction by p53 class mediator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C5orf35
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SETD9
Gene Accession No. Q8NE22
Gene Alias C5orf35; SET domain containing 9; SET domain-containing protein 9; SETD9
Gene Symbols SETD9
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human C5orf35.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133383
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.