missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: IAENIEQDFP KPEMLDLHCD KIGPVDHIEF SSGPEFKKTL ASKTISISEE VRFLTHMDEE DQSDKKETSE PEFSITE Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 29%.
Specifications
Specifications
| Antigen | C5orf42 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | C5orf42 |
| Gene Accession No. | Q9H799 |
| Gene Alias | C5orf42; chromosome 5 open reading frame 42; Ciliogenesis and planar polarity effector 1; CPLANE1; Hug; JBTS17; OFD6; Protein JBTS17; Transmembrane protein ENSP00000382582; uncharacterized protein C5orf42 |
| Gene Symbols | C5orf42 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
