Learn More
BACE-1 Antibody (137612) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB931B
Product Code. 18663715
  • ONLINE EXCLUSIVE PRICE
    € 706.00
    € 667.80 / 0.10mL
    Save € 38.20
    5% Off
Description
BACE-1 Monoclonal antibody specifically detects BACE-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BACE-1
Monoclonal
Biotin
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human, Mouse
Purified
Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
137612
Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2
Mouse myeloma cell line NS0-derived recombinant human BACE-1, Thr22-Tyr460, Accession # P56817
0.1 mL
Alzheimers Research, Neurodegeneration, Neuroscience
23621
Store at 4°C in the dark.