missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5orf49 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 307.65
Specifications
| Antigen | C5orf49 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18675322
|
Novus Biologicals
NBP2-98031-100ul |
100 μg |
€ 326.00 € 307.65 / 100µL Save € 18.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663714
|
Novus Biologicals
NBP2-98031-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C5orf49 Polyclonal antibody specifically detects C5orf49 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| C5orf49 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C5orf49 chromosome 5 open reading frame 49 | |
| Produced in rabbits immunized with E. coli-derived Human C5orf49 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 134121 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title