missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C5orf54 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA561001

Product Code. 15708821

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: NQFLHHKSSN LVDGFENKEF KIHLAYLADL FKHLNELSAS MQRTGMNTVS AREKLSAFVR KFPFWQKRIE KRNFTNFPFL EEIIVSDNEG IFIAAEITLH L Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 33%.

ZBED8 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C5orf54
Polyclonal
Unconjugated
ZBED8
Buster3; C5orf54; Protein ZBED8; transposon-derived Buster3 transposase-like protein; ZBED8; Zinc finger BED domain-containing protein 8; zinc finger BED-type containing 8; zinc finger, BED-type containing 8
Rabbit
Antigen affinity chromatography
RUO
63920
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
Lot-Specific
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IZ13
ZBED8
Recombinant protein corresponding to Human C5orf54. Recombinant protein control fragment (Product #RP-100354).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.