Learn More
C6 Mouse anti-Human, Clone: OTI1H4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF812708
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
C6 is a component of complement cascade. It is part of the membrane attack complex which can insert into the cell membrane and cause cell to lyse. People with C6 deficiency are prone to bacterial infection. Two transcript variants encoding the same protein have been found for this gene.Specifications
| C6 | |
| Monoclonal | |
| Unconjugated | |
| C6 | |
| complement C6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 729 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1H4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P13671 | |
| C6 | |
| Human recombit protein fragment corresponding to amino acids 176-522 of human C6 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.