missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C6orf211 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA551793

Product Code. 15719750

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VEAEKKAISL LSKLRNELQT DKPFIPLVEK FVDTDIWNQY LEYQQSLLNE SDGKSRWFYS PWLLVECYMY RRIHEAIIQS PPIDYFDVFK ESKEQNFYGS QESIIALCTH LQQLIRTIED L Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 81%.

ARMT1 gene ontology annotations related to this gene include protein carboxyl O-methyltransferase activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C6orf211
Polyclonal
Unconjugated
ARMT1
Acidic residue methyltransferase 1; ARMT1; C6orf211; Damage-control phosphatase ARMT1; Protein-glutamate O-methyltransferase; Sugar phosphate phosphatase ARMT1; UPF0364 protein C6orf211
Rabbit
Antigen Affinity Chromatography
RUO
79624
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H993
ARMT1
Recombinant protein corresponding to Human C6orf211. Recombinant protein control fragment (Product #RP-102001).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.