Learn More
C6orf211 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25124-1-AP-20UL
 View more versions of this product
Product Code. 16803675
  • € 145.00 / 20µL
Description
ARMT1 gene ontology annotations related to this gene include protein carboxyl O-methyltransferase activity.
Specifications
Specifications
C6orf211
Polyclonal
Unconjugated
ARMT1
C6orf211, UPF0364 protein C6orf211
Rabbit
Antigen Affinity Chromatography
RUO
79624
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H993
ARMT1
C6orf211 Fusion Protein Ag19090