Learn More
C6orf47 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98022MFV500
This item is not returnable.
View return policy
Description
C6orf47 Polyclonal antibody specifically detects C6orf47 in Human samples. It is validated for Immunohistochemistry (Paraffin)