missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C6orf64 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569471
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
C6orf64 (SAYSD1, SAYSVFN Motif Domain Containing 1) is a Protein Coding gene.
Specifications
| C6orf64 | |
| Polyclonal | |
| Unconjugated | |
| SAYSD1 | |
| 1810063B07Rik; 4930488P03Rik; C12H6orf64; C23H6orf64; C6orf64; SAYSD1; SAYSvFN domain-containing protein 1; SAYSVFN motif domain containing 1; uncharacterized protein C6orf64 homolog; Unknown (protein for MGC:134290) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55776 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NPB0 | |
| SAYSD1 | |
| synthetic peptide directed towards the C terminal region of human C6orf64. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction