missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C6orf81, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00221481-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MGKSIPQYLGQLDIRKSVVSLATGAGAIYLLYKAIKAGIKCKPPLCSNSPICIARECPGPGERALPQEAPAPEASAVGGPKGLAVERERHGRDSGELRRLLNSLECKQDEYAKSMILHSITRCVYLLEAEASACTTDDIVLLGYMLDDKDNSVKTQALNTLKAFSGIRKFRLKIQEHSIKVLELISTIWDTELHIAGLRLLNNLPLPDYVHPQLRRVMPALMEILQSDYILAQVQAVRLLSYLAQKNDLLYDILNCQVHSNFLNLFQPTQSGSLLYEVLVFAERLSEGRNAPHYHVVKWHYNEQSLHESLFGEESRLADRLLALVIHPEEDVQIQACKVIVSLQYPQDLRARPSSCQPSRSYFKNTESpecifications
| C6orf81 | |
| Polyclonal | |
| No additive | |
| NM_145028 | |
| C6orf81 | |
| C6orf81 (NP_659465, 1 a.a. ∼ 367 a.a) full-length human protein. | |
| RUO | |
| 221481 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| chromosome 6 open reading frame 81 | |
| FLJ25390 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction