missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C6orf89 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144145
This item is not returnable.
View return policy
Description
C6orf89 Polyclonal Antibody for IHC (P), ELISA
Specifications
| C6orf89 | |
| Polyclonal | |
| Unconjugated | |
| C6orf89 | |
| amfion; Bombesin receptor-activated protein C6orf89; BRAP; C6orf89; chromosome 6 open reading frame 89; UNQ177/PRO203 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 221477 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q6UWU4 | |
| C6orf89 | |
| Recombinant Human Bombesin receptor-activated protein C6orf89 protein (120-320AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction