missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C6orf97 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA582953

Product Code. 16329914

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RMVSQLEAQI SELVEQLGKE SGFHQKALQR AQKAENMLET LQGQLTHLEA ELVSGGVLRD NLNFEKQKYL KFLDQLSQKM KLDQMA Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 76%.

The function of this gene and its encoded protein is not known. Several genome-wide association studies have implicated the region around this gene to be involved in breast cancer and bone mineral density, but no link to this specific gene has been found. [provided by RefSeq, May 2010].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C6orf97
Polyclonal
Unconjugated
CCDC170
bA282P11.1; C6orf97; CCDC170; cf097; coiled-coil domain containing 170; coiled-coil domain-containing protein 170; Gm221; RGD1310945
Rabbit
Antigen affinity chromatography
RUO
80129
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IYT3
CCDC170
Recombinant protein corresponding to Human CCDC170. Recombinant protein control fragment (Product #RP-102546).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.