missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C7orf20 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17147693
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17147693 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17147693 Supplier Invitrogen™ Supplier No. PA5109762

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MAAAAAMAEQ ESARNGGRNR GGVQRVEGKL RASVEKGDYY EAHQMYRTLF FRYMSQSKHT EARELMYSGA LLFFSHGQQN SAADLSMLVL ESLEKAEVEV ADELLENLAK VFSLMDPNSP ERVTFVSRAL KWSSGGSGKL GHPRLHQLLA LTLWKEQNYC ESRYHFLHSA DGEGCANMLV EYSTSRGFRS EVDMFVAQAV LQFLCLKNKS SASVVFTTYT QKHPSIEDGP PFVEPLLNFI WFLLLAVDGG KLTVFTVLCE QYQPSLRRDP MYNEYLDRIG QLFFGVPPKQ TSSYGGLLGN LLTSLMGSSE QEDGEESPSD GSPIELD.

As part of a cytosolic protein quality control complex, the BAG6/BAT3 complex, maintains misfolded and hydrophobic patches-containing proteins in a soluble state and participates in their proper delivery to the endoplasmic reticulum or alternatively can promote their sorting to the proteasome where they undergo degradation (PubMed:20676083, PubMed:21636303, PubMed:21743475, PubMed:28104892). The BAG6/BAT3 complex is involved in the post-translational delivery of tail-anchored/type II transmembrane proteins to the endoplasmic reticulum membrane. Recruited to ribosomes, it interacts with the transmembrane region of newly synthesized tail-anchored proteins and together with SGTA and ASNA1 mediates their delivery to the endoplasmic reticulum (PubMed:20676083, PubMed:28104892, PubMed:25535373). Client proteins that cannot be properly delivered to the endoplasmic reticulum are ubiquitinated and sorted to the proteasome (PubMed:28104892). Similarly, the BAG6/BAT3 complex also functions as a sorting platform for proteins of the secretory pathway that are mislocalized to the cytosol either delivering them to the proteasome for degradation or to the endoplasmic reticulum (PubMed:21743475). The BAG6/BAT3 complex also plays a role in the endoplasmic reticulum-associated degradation (ERAD), a quality control mechanism that eliminates unwanted proteins of the endoplasmic reticulum through their retrotranslocation to the cytosol and their targeting to the proteasome. It maintains these retrotranslocated proteins in an unfolded yet soluble state condition in the cytosol to ensure their proper delivery to the proteasome (PubMed:21636303). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C7orf20
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GET4
Gene Accession No. Q7L5D6, Q9D1H7
Gene Alias 1110007L15Rik; AW412535; C7orf20; CEE; CGI-20; conserved edge expressed protein; conserved edge protein; conserved edge-expressed protein; GET4; golgi to ER traffic protein 4; Golgi to ER traffic protein 4 homolog; golgi to ER traffic protein 4 homolog (S. cerevisiae); H_NH1244M04.5; RGD1311660; Transmembrane domain recognition complex 35 kDa subunit; transmembrane domain recognition complex, 35kDa; TRC35
Gene Symbols GET4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-327 of human GET4 (NP_057033.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288518, 51608, 67604
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.