missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C7orf25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565571
This item is not returnable.
View return policy
Description
Immunogen sequence: RVDRENILASV AFPTEIKVDV CKRVNLDITT LITYVSALSY GGCHFIFKEK VLTEQAEQER KEQVLPQLEA FMKDKELFAC ESAVKDF Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
C7orf25 is a protein coding gene.
Specifications
| C7orf25 | |
| Polyclonal | |
| Unconjugated | |
| C7orf25 | |
| C7orf25; chromosome 7 open reading frame 25; UPF0415 protein; UPF0415 protein C7orf25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79020 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BPX7 | |
| C7orf25 | |
| Recombinant Human C7orf25. Recombinant protein control fragment (Product #RP-104878). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction