Learn More
Invitrogen™ C7orf31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113207
This item is not returnable.
View return policy
Description
C7orf31 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
C7orf31 is a protein coding gene.
Specifications
| C7orf31 | |
| Polyclonal | |
| Unconjugated | |
| C7orf31 | |
| C7orf31; chromosome 7 open reading frame 31; uncharacterized protein C7orf31 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 136895 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8N865 | |
| C7orf31 | |
| Recombinant Human Uncharacterized protein C7orf31 protein (163-256AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |