Learn More
Invitrogen™ C7orf38 Monoclonal Antibody (4E4)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA524556
Product Code. 15642385
  • € 534.00 / 100µg
Description
Peptide Sequence: MTPESRDTTD LSPGGTQEME GIVIVKVEEE DEEDHFQKER NKVESSPQVL SRSTTMNERA LLSSYLVAYR VAKEKMAHTA AEKIILPACM DMVRTIFDDK SADKLRTIPL SDNTISRRIC TIAKHLEAML ITRLQSGIDF AIQLDESTDI ASCPTLLVYV RYVWQDDFVE DLLCCLNLNS HITGLDLFTE LENCLLGQYK LNWKHCKGIS SDGTANMTGK HSRLTEKLLE ATHNNAVWNH CFIHREALVS KEISPSLMDV LKNAVKTVNF IKGSSLNSRL LEIFCSEIGV NHTHLLFHTE VRWLSQGKVL SRVYELRNEI YIFLVEKQSH LANIFEDDIW VTKLAYLSDI FGILNELSLK MQGKNNDIFQ YLEHILGFQK TLLLWQARLK SNRPSYYMFP TLLQHIEENI INEDCLKEIK LEILLHLTSL SQTFNYYFPE EKFESLKENI WMKDPFAFQN PESIIELNLE PEEENELLQL SSSFTLKNYY KILSLSAFWI KIKDDFPLLS RKSILLLLPF TTTYLCELGF SILTRLKTKK RNRLNSAPDM RVALSSCVPD WKELMNRQAH PSH.
This gene encodes a protein of unknown function. The protein is weakly similar to transposase-like proteins in human and mouse.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C7orf38
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
Q8TCP9
FAM200A
C7orf38 (AAH40214, 1 a.a. ~ 573 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2b κ
ELISA, Western Blot
4E4
Unconjugated
FAM200A
C7orf38; FAM200A; family with sequence similarity 200 member A; family with sequence similarity 200, member A; protein FAM200A
Mouse
Protein A
RUO
221786
-20°C, Avoid Freeze/Thaw Cycles