missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C7orf38 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553644
This item is not returnable.
View return policy
Description
Immunogen sequence: ERALLSSYLV AYRVAKEKMA HTAAEKIILP ACMDMVRTIF DDKSADKLRT IPLSDNTISR RICTIAKHLE AMLITRLQSG IDFAIQLDES TDIASCPTLL VYVRYVWQDD FVEDLLCCLN LNSHITGLDL FTELENCLLG QYKLN Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 39%.
This gene encodes a protein of unknown function. The protein is weakly similar to transposase-like proteins in human and mouse.
Specifications
| C7orf38 | |
| Polyclonal | |
| Unconjugated | |
| FAM200A | |
| C7orf38; FAM200A; family with sequence similarity 200 member A; family with sequence similarity 200, member A; protein FAM200A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 221786 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TCP9 | |
| FAM200A | |
| Recombinant protein corresponding to Human C7orf38. Recombinant protein control fragment (Product #RP-90556). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction