missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C8G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145304
This item is not returnable.
View return policy
Description
C8G is one of the three polypeptides that constitute C8, a component of the complement system. C8 participates in the formation of Membrane Attack Complex (MAC). Patients with deficiency in C8 are vulnerable to certain bacteria infection.
Specifications
| C8G | |
| Polyclonal | |
| Unconjugated | |
| C8G | |
| C8C; C8G; complement component 8, gamma polypeptide; complement component C8 gamma chain | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100037955, 733 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P07360 | |
| C8G | |
| Recombinant Human C8g protein, Lys39-Arg202 (Accession #P07360). | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction