missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C9orf126 Monoclonal Antibody (7-A4)
GREENER_CHOICE

Product Code. 16336284
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16336284 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16336284 Supplier Invitrogen™ Supplier No. MA532873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

C9orf126 Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

This gene encodes a regulator of cell migration. The encoded protein appears to function in the RhoA (ras homolog gene family, member A)-Dia1 (diaphanous homolog 1) signal transduction pathway. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf126
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7-A4
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene SCAI
Gene Accession No. Q8N9R8
Gene Alias 9330112M16; A930041I02Rik; AI662729; C9orf126; NET40; protein SCAI; SCAI; suppressor of cancer cell invasion; suppressor of cancer cell invasion protein
Gene Symbols SCAI
Host Species Mouse
Immunogen Recombinant protein within Human SCAI aa 1-606.
Purification Method Protein L
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286205
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.