missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AJM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554349
This item is not returnable.
View return policy
Description
Immunogen sequence: PRPYPSEELS GPSPRRMGGY YAGEVRTFPI QEPPSRSYYG EAPRAYGLPY GPRYVPEEPR AHSTARPFYT EDFGRYRERD VLARTYPHPR SSPAW Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.
AJM1 is a transmembrane protein that plays a crucial role in cell-cell adhesion and maintenance of epithelial cell polarity. It is found in the junctional complex of epithelial cells and is involved in the formation and stabilization of tight junctions.
Specifications
| AJM1 | |
| Polyclonal | |
| Unconjugated | |
| AJM1 | |
| AJM1; Apical junction component 1 homolog; C9orf172; chromosome 9 open reading frame 172; uncharacterized protein C9orf172 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 389813 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| C9J069 | |
| AJM1 | |
| Recombinant protein corresponding to Human C9orf172. Recombinant protein control fragment (Product #RP-93082). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction