missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C9orf24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570776
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 86%; Rat: 93%.
This gene encodes a nuclear- or perinuclear-localized protein with no predicted domains or similarity to other known proteins. Expression of this gene is induced during the differentiation of bronchial epithelial cells, and the encoded protein may play a role in ciliogenesis. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| C9orf24 | |
| Polyclonal | |
| Unconjugated | |
| C9orf24 | |
| bA573M23.4; C9orf24; CBE1; chromosome 9 open reading frame 24; ciliated bronchial epithelial protein 1; ciliated bronchial epithelium 1; NYD-SP22; SMRP1; Spermatid-specific manchette-related protein 1; testis development protein NYD-SP22 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84688 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NCR6 | |
| C9orf24 | |
| synthetic peptide directed towards the N-terminal region of human C9orf24. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction