missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf41 Monoclonal Antibody (OTI1E8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501299S
This item is not returnable.
View return policy
Description
C9orf41 Monoclonal antibody specifically detects C9orf41 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| C9orf41 | |
| Monoclonal | |
| 0.89 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5BJZ6, Q8N4J0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 138199, 293871, 476320 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1E8 | |
| Unconjugated | |
| CARNMT1 | |
| 2410127L17Rik; Anserine-producing methyltransferase; C1H9orf41; C9orf41; CARNMT1; Carnosine N-methyltransferase; carnosine N-methyltransferase 1; RGD1311863; RIKEN cDNA 2410127L17 gene; similar to RIKEN cDNA 2410127L17; UPF0586; UPF0586 protein C9orf41; UPF0586 protein C9orf41 homolog | |
| Full length human recombinant protein of human C9orf41 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction