Learn More
C9orf41 Mouse anti-Human, Clone: OTI4B7, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501308
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
N-methyltransferase that mediates the formation of anserine (beta-alanyl-N(Pi)-methyl-L-histidine) from carnosine. Anserine, a methylated derivative of carnosine (beta-alanyl-L-histidine), is an abundant constituent of vertebrate skeletal muscles. Also methylates other L-histidine-containing di- and tripeptides such as Gly-Gly-His, Gly-His and homocarnosine (GABA-His).Specifications
| C9orf41 | |
| Monoclonal | |
| Unconjugated | |
| CARNMT1 | |
| C9orf41, UPF0586 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 138199 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B7 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8N4J0 | |
| CARNMT1 | |
| Full length human recombit protein of human C9orf41 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.