missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf72 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82070AF750
This item is not returnable.
View return policy
Description
C9orf72 Polyclonal antibody specifically detects C9orf72 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| C9orf72 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 750 | |
| ALSFTD, chromosome 9 open reading frame 72, FTDALS | |
| C9orf72 antibody was raised against a 19 amino acid peptide from near the carboxy terminus of human C9orf72. The immunogen is located within amino acids 400 - 450 of C9orf72. Amino Acid Squence: KYIEDDTQKGKKPFKSLRN | |
| 0.1 mL | |
| Neuroscience | |
| 203228 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction