missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf72 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47146H
This item is not returnable.
View return policy
Description
C9orf72 Polyclonal specifically detects C9orf72 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| C9orf72 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ALSFTD, chromosome 9 open reading frame 72, FTDALS | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS with No Preservative | |
| C9orf72 | |
| A synthetic peptide made to an internal sequence of the human C9orf72 protein [UniProt Q96LT7]. | |
| 0.1 mL | |
| Neuroscience | |
| 203228 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction