Learn More
C9orf72 Mouse anti-Human, Mouse, Rat, Clone: 3D2H6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66140-1-IG-150UL
This item is not returnable.
View return policy
Description
C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis.Specifications
| C9orf72 | |
| Monoclonal | |
| 1.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96LT7 | |
| C9orf72 | |
| C9orf72 Fusion Protein Ag21080 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 3D2H6 | |
| Unconjugated | |
| C9orf72 | |
| C9RANT | |
| Mouse | |
| Protein A | |
| RUO | |
| 203228, 313155, 73205 | |
| -20°C | |
| Liquid |