missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf72 Mouse anti-Human, Mouse, Rat, Clone: 3D2H6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66140-1-IG-20UL
This item is not returnable.
View return policy
Description
C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis.Specifications
| C9orf72 | |
| Monoclonal | |
| 1.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96LT7 | |
| C9orf72 | |
| C9orf72 Fusion Protein Ag21080 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG2a |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
| 3D2H6 | |
| Unconjugated | |
| C9orf72 | |
| C9RANT | |
| Mouse | |
| Protein A | |
| RUO | |
| 203228, 313155, 73205 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction