missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf72 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25757-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody detects the N-terminal of C9orf72.
C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis.Specifications
| C9orf72 | |
| Polyclonal | |
| Unconjugated | |
| C9orf72 | |
| C9orf72, C9RANT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 203228, 73205 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96LT7 | |
| C9orf72 | |
| C9orf72 Fusion Protein Ag22723 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction