missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: MPVVRKIFRR RRGDSESEED EQDSEEVRLK LEETREVQNL RKRPNGVSAV ALLVGEKVQE ETTLVDDPFQ MKTGGMVDMK KLKE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Involved in the regulation of telomeric heterochromatin assembly and control of telomere length. [UniProt].
Specifications
Specifications
| Antigen | C9orf78 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | C9orf78 |
| Gene Accession No. | Q9NZ63 |
| Gene Alias | bA409K20.3; C9orf78; chromosome 9 open reading frame 78; HCA59; Hepatocellular carcinoma-associated antigen 59; HSPC220; RGD1305178; similar to Hypothetical protein MGC11690; Telomere length and silencing protein 1 homolog; uncharacterized protein C9orf78; uncharacterized protein C9orf78 homolog |
| Gene Symbols | C9orf78, RGD1305178 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
