missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf78 Polyclonal Antibody

C9orf78 Polyclonal Antibody
Brand: Thermo Scientific PA554356
This item is not returnable.
View return policy
Description
C9orf78 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| C9orf78 | |
| Polyclonal | |
| Unconjugated | |
| C9orf78 | |
| bA409K20.3; C9orf78; chromosome 9 open reading frame 78; HCA59; Hepatocellular carcinoma-associated antigen 59; HSPC220; RGD1305178; similar to Hypothetical protein MGC11690; Telomere length and silencing protein 1 homolog; uncharacterized protein C9orf78; uncharacterized protein C9orf78 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51759 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZ63 | |
| C9orf78 | |
| Recombinant protein corresponding to Human C9orf78. Recombinant protein control fragment (Product #RP-92960). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction