missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C9orf78 Polyclonal Antibody

Product Code. 13279838
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13279838 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13279838 Supplier Invitrogen™ Supplier No. PA521806

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Xenopus laevis (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Involved in the regulation of telomeric heterochromatin assembly and control of telomere length. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf78
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene C9orf78
Gene Accession No. Q9NZ63
Gene Alias bA409K20.3; C9orf78; chromosome 9 open reading frame 78; HCA59; Hepatocellular carcinoma-associated antigen 59; HSPC220; RGD1305178; similar to Hypothetical protein MGC11690; Telomere length and silencing protein 1 homolog; uncharacterized protein C9orf78; uncharacterized protein C9orf78 homolog
Gene Symbols C9orf78, RGD1305178
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human C9orf78. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311855, 51759
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.