missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf89 Monoclonal Antibody (OTI2B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811304
This item is not returnable.
View return policy
Description
Plays a role in inhibiting the effects of BCL10-induced activation of NF-kappa-B. May inhibit the phosphorylation of BCL10 in a CARD-dependent manner.Specifications
| C9orf89 | |
| Monoclonal | |
| Unconjugated | |
| CARD19 | |
| BinCARD, C9orf89 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84270 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2B12 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96LW7 | |
| CARD19 | |
| Full length human Recombit protein of human C9ORF89 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction