missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf89 Monoclonal Antibody (OTI2B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811304S
This item is not returnable.
View return policy
Description
C9orf89 Monoclonal antibody specifically detects C9orf89 in Human samples. It is validated for Western BlotSpecifications
| C9orf89 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11217 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5837 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI2B12 | |
| Unconjugated | |
| PYGM | |
| 1110007C09Rik; AI851695; Bcl10-interacting CARD protein; Bcl10-interacting protein with CARD; BinCARD; C9orf89; CARD19; caspase recruitment domain family member 19; caspase recruitment domain family, member 19; caspase recruitment domain-containing protein 19; RIKEN cDNA 1110007C09 gene | |
| Human recombinant protein fragment corresponding to amino acids 698-842 of human PYGM produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction