missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA125/MUC16 Antibody (CL2783) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59024R
This item is not returnable.
View return policy
Description
CA125/MUC16 Monoclonal antibody specifically detects CA125/MUC16 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| CA125/MUC16 | |
| Monoclonal | |
| DyLight 550 | |
| CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
| This antibody was developed using a recombinant protein derived from Q8WXI7, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers | |
| 94025 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL2783 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction