missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA125/MUC16 Antibody (OTI2C4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72336B
This item is not returnable.
View return policy
Description
CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CA125/MUC16 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| MUC16 | |
| Recombinant protein fragment corresponding to amino acids 10329-10628 of human CA125/MUC16 (NP_078966) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2C4 | |
| Western Blot, Immunohistochemistry | |
| CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers | |
| 94025.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction