missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA125/MUC16 Antibody (OTI3C6), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72337APC
This item is not returnable.
View return policy
Description
CA125/MUC16 Monoclonal antibody specifically detects CA125/MUC16 in Human samples. It is validated for Western Blot
Specifications
| CA125/MUC16 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| OTI3C6 | |
| Western Blot | |
| CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
| Human recombinant protein fragment corresponding to amino acids 13942-14063 and 21117-21238 of human MUC16 (NP_078966) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers | |
| 94025 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction