missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CA14 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17121374 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17121374 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17121374 Supplier Invitrogen™ Supplier No. PA5113512

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CA14 Polyclonal Antibody for Western Blot, IHC (P), ELISA

CA14 belongs a large family of zinc metalloenzymes that catalyze the versible hydration of carbon dioxide. The protein is predicted to be a type I membrane protein and shares highest sequence similarity with the other transmembrane CA isoform, CA XII; however, they have different patterns of tissue-specific expression and thus may play different physiologic roles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CA14
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CA14
Gene Accession No. Q9ULX7, Q9WVT6
Gene Alias AW536446; CA XIV; CA14; Car14; Carbonate dehydratase XIV; carbonic anhydrase 14; Carbonic anhydrase XIV; carbonic dehydratase; Catm; CAXiV; CA-XIV; UNQ690/PRO1335
Gene Symbols CA14, Car14
Host Species Rabbit
Immunogen Recombinant Human Carbonic anhydrase 14 protein (16-290AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23632, 23831
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.