Learn More
CA19-9 Mouse anti-Human, Clone: NS19-9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF190083
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mucin glycoprotein is a Sialyl Lewis A structure which is synthesized from type 1 blood group precursor chains and is present in individuals expressing the Lewis A and/or Lewis B blood group antigens. In normal tissues, sialyl Lewis A antigen is present in ductal epithelium of the breast, kidney, salivary gland, and sweat glands. Its expression is greatly enhanced in serum as well as in the majority of tumor cells in gastrointestinal (GI) carcinomas, including adenocarcinomas of the stomach, intestine, and pancreas. Preoperative elevated CA19-9 levels in patients with stage I pancreatic carcinoma decrease to normal values following surgery. When used serially, CA19-9 can predict recurrence of disease prior to radiographic or clinical findings.Specifications
| CA19-9 | |
| Monoclonal | |
| Unconjugated | |
| CA19-9 | |
| SW1116 human colon carcinoma cell line | |
| 100 μg | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| NS19-9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.