Learn More
Kappes Systeme™ Cabinet
Click to view available options
Color:
Light Gray
Material:
Galvanized Steel Decks, Metal Doors
Unit Size:
Each
Specifications
Specifications
| Product Type | System Cabinet |
| Color | Light Gray |
| Cabinet Type | Yes |
| Depth (Metric) | 410 mm |
| Dimensions (D x W x H) | 410 x 1000 x 1950 mm |
| Height (Metric) | 195 cm |
| Material | Galvanized Steel Decks, Metal Doors |