missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX-1/OLR1 Antibody (331212), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1798B
This item is not returnable.
View return policy
Description
LOX-1/OLR1 Monoclonal antibody specifically detects LOX-1/OLR1 in Human samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Specifications
| LOX-1/OLR1 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 331212 | |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX-1, LOX1ox LDL receptor 1, LOXIN, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1 | |
| Mouse myeloma cell line NS0-derived recombinant human LOX-1/OLR1, Ser61-Gln273, Accession # P78380 | |
| 0.1 mL | |
| Cardiovascular Biology, metabolism | |
| 4973 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction