missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CABLES1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 193.00 - € 463.00
Specifications
| Antigen | CABLES1 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695662
|
Novus Biologicals
NBP2-92697-0.02ml |
0.02 mL |
€ 193.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671082
|
Novus Biologicals
NBP2-92697-0.1ml |
0.1 mL |
€ 463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CABLES1 Polyclonal antibody specifically detects CABLES1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| CABLES1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication | |
| PBS with 50% glycerol, pH7.3. | |
| 91768 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CABL1, CABLES, Cdk5 and Abl enzyme substrate 1, CDK5 and ABL1 enzyme substrate 1, FLJ35924, HsT2563, IK3-1, Interactor with CDK3 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-368 of human CABLES1 (NP_612384.1). MLSKRGCHARIYADFPIRRLISQRSSLETLEDIEENAPLRRCRTLSGSPRPKNFKKIHFIKNMRQHDTRNGRIVLISGRRSFCSIFSVLPYRDSTQVGDLKLDGGRQSTGAVSLKEIIGLEGVELGADGKTVSYTQFLLPTNAFGARRNTIDSTSSFSQFRNLSHRSLSIGRASGTQGSLDTGSDLGDFMDYDPNLLDDPQWPCGKHKRVLIFPSYMTTVIDYVKPSDLKKDMNETFKEKFPHIKLTLSKIRSLKREMRKLAQEDCGLEEPTVAMAFVYFEKLALKGKLNKQNRKLCAGACVLLAAKIGSDLKKHEVKHLIDKLEEKFRLNRRELIAFEFPVLVALEFALHLPEHEVMPHYRRLVQSS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit