missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CABLES1 Monoclonal Antibody (OTI2A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811422S
This item is not returnable.
View return policy
Description
CABLES1 Monoclonal antibody specifically detects CABLES1 in Canine, Human, Rat samples. It is validated for Western BlotSpecifications
| CABLES1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| B0K035, Q6P444 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 100911069, 113115, 483997 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2A3 | |
| Unconjugated | |
| MTFR2 | |
| CABL1; CABLES; CABLES1; cdk3-binding protein ik3-1; CDK5 and Abl enzyme substrate 1; CDK5 and ABL1 enzyme substrate 1; HsT2563; ik3-1; interactor with CDK3 1; interactor-1 with cdk3 | |
| Full length human recombinant protein of human FAM54A produced in E.coli. | |
| 30 μL | |
| Primary | |
| Canine, Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction