missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CABP4 Antibody (OTI9A7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72071F
This item is not returnable.
View return policy
Description
CABP4 Monoclonal antibody specifically detects CABP4 in Human samples. It is validated for Western Blot
Specifications
| CABP4 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 57010 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot | |
| OTI9A7 | |
| Western Blot | |
| CaBP4, calcium binding protein 4, CSNB2Bcalcium-binding protein 4 | |
| Full length human recombinant protein of human CABP4(NP_660201) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction