missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CABP5 Antibody (08), DyLight 488 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06210G
This item is not returnable.
View return policy
Description
CABP5 Monoclonal antibody specifically detects CABP5 in Human samples. It is validated for ELISA
Specifications
| CABP5 | |
| Monoclonal | |
| DyLight 488 | |
| CABP3, calcium binding protein 5, calcium-binding protein 3, calcium-binding protein 5 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CABP5 (Q9NP86; Met1-Arg173). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 08 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 56344 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction