missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CABP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586759
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
The calcium binding protein (CaBP) family shares much similarity to calmodulin. It has been shown that CaBP proteins can substitute functionally for, and probably augment the function of, calmodulin. Calcium binding proteins are a crucial part of calcium mediated cellular signal transduction in the central nervous system. There are several members of the family with varying expression patterns. CaBP1 and CaBP2 can be expressed as multiple, alternatively spliced variants in brain and retina. CaBP3, CaBP4 and CaBP 5 are restricted to retinal rod and cone cells.
Specifications
| CABP5 | |
| Polyclonal | |
| Unconjugated | |
| CABP5 | |
| CABP3; CaBP5; calcium binding protein 5; calcium-binding protein 3; calcium-binding protein 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29865, 365194, 56344 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9JLK3, Q9NP86 | |
| CABP5 | |
| A synthetic peptide corresponding to residues in Human CABP5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction